missing translation for 'onlineSavingsMsg'
Learn More

Neophytadiene (80%), TRC

Product Code. 30004898
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004898 100 mg 100mg
17968304 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004898 Supplier Toronto Research Chemicals Supplier No. TRCN390000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-96-1
Molecular Formula C20 H38
Molecular Weight (g/mol) 278.52
Synonym 7,11,15-Trimethyl-3-methylene-1-hexadecene, 2-(4,8,12-Trimethyltridecyl)-1,3-butadiene, 2-(4,8,12-Trimethyltridecyl)buta-1,3-diene, 3-Methylene-7,11,15-trimethylhexadec-1-ene;
IUPAC Name 7,11,15-trimethyl-3-methylidenehexadec-1-ene
SMILES CC(C)CCCC(C)CCCC(C)CCCC(=C)C=C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H38/c1-7-18(4)12-9-14-20(6)16-10-15-19(5)13-8-11-17(2)3/h7,17,19-20H,1,4,8-16H2,2-3,5-6H3
Formula Weight 278.2974
Chemical Name or Material Neophytadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.