missing translation for 'onlineSavingsMsg'
Learn More

Neophytadiene (80%) (1mg/mL in Chloroform), TRC

Product Code. 30467325
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467325 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467325 Supplier Toronto Research Chemicals Supplier No. TRCKIT96401X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 504-96-1
Molecular Formula C20 H38
Quantity 1 mL
Synonym 7,11,15-Trimethyl-3-methylene-1-hexadecene, 2-(4,8,12-Trimethyltridecyl)-1,3-butadiene, 2-(4,8,12-Trimethyltridecyl)buta-1,3-diene, 3-Methylene-7,11,15-trimethylhexadec-1-ene
InChI Formula InChI=1S/C20H38/c1-7-18(4)12-9-14-20(6)16-10-15-19(5)13-8-11-17(2)3/h7,17,19-20H,1,4,8-16H2,2-3,5-6H3
SMILES CC(C)CCCC(C)CCCC(C)CCCC(=C)C=C
IUPAC Name 7,11,15-trimethyl-3-methylidenehexadec-1-ene
Molecular Weight (g/mol) 278.52
Formula Weight 278.2974
Concentration or Composition (by Analyte or Components) Neophytadiene 1 mg/mL
Chemical Name or Material Neophytadiene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.