accessibility menu, dialog, popup

UserName

NH2N

CAS RN 504-24-5

IUPAC Name: pyridin-4-amine
Synonyms: 4 aminopyridine epa pesticide chemical code 069201 caswell no. 038 4-aminopyridine (substance) 4-aminopyridine (product) 氨吡啶 amino-4 pyridine avitrol 200 compound 1861 4 aminopyridine sustained release
Molecular Weight (g/mol): 94.12
Molecular Formula: C5H6N2
InChi Key: NUKYPUAOHBNCPY-UHFFFAOYSA-N
SMILES: NC1=CC=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (7)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (7)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (5)
  • (1)
  • (2)
  • (3)
  • (7)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (5)
  • (2)
  • (7)
  • (4)
  • (2)