accessibility menu, dialog, popup

UserName

H2COOHOPOOHOK

CAS RN 4265-07-0

IUPAC Name: potassium 2-(hydrogen phosphonatooxy)prop-2-enoic acid
Synonyms: potassium 2-(hydrogen phosphonatooxy)prop-2-enoic acid
Molecular Weight (g/mol): 206.13
Molecular Formula: C3H4KO6P
InChi Key: SOSDSEAIODNVPX-UHFFFAOYSA-M
SMILES: [K+].OC(=O)C(=C)OP(O)([O-])=O
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (1)
  • (8)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)