missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic acid, monopotassium salt, MP Biomedicals™

Product Code. 11492511 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
0.1 g
250 mg
1 g
Unit Size:
100mg
1g
250mg
25mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492511 250 mg 25mg
11412521 25 mg 250mg
11402521 0.1 g 100mg
11422521 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11492511 Supplier MP Biomedicals Supplier No. 0219570725
  • ONLINE EXCLUSIVE PRICE
    44.10 EUR
    42.00 EUR / 25mg
    Save 2.10 EUR
    4% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4265-07-0
Molecular Formula C3H4KO6P
Molecular Weight (g/mol) 206.131
MDL Number MFCD00044476
InChI Key SOSDSEAIODNVPX-UHFFFAOYSA-M
Synonym potassium 1-carboxyvinyl hydrogenphosphate, potassium hydrogen 2-phosphonatooxy acrylate, 2-propenoic acid, 2-phosphonooxy-, monopotassium salt, potassium 1-carboxyvinyl hydrogen phosphate, phospho enol pyruvic acid monopotassium salt, phosphoenolpyruvic acid monopotassium salt, 2-propenoic acid, 2-phosphonooxy-, potassium salt 1:1, potassium 2-hydrogen phosphonooxy prop-2-enoic acid, phosphoenolpyruvate potassium salt, potassium 2-hydrogen phosphonatooxy prop-2-enoic acid
PubChem CID 23678879
IUPAC Name potassium;1-carboxyethenyl hydrogen phosphate
SMILES C=C(C(=O)O)OP(=O)(O)[O-].[K+]

Specifications

Color White
Quantity 250 mg
Formula Weight 206.1
Physical Form Crystalline Powder
Chemical Name or Material Phosphoenolpyruvic acid, monopotassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.