missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic Acid Monopotassium Salt, MP Biomedicals™

Product Code. 11468312 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468312 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11468312 Supplier MP Biomedicals Supplier No. 0219570794
  • ONLINE EXCLUSIVE PRICE
    1660.00 EUR
    1577.10 EUR / 5g
    Save 82.90 EUR
    4% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Functions as an important metabolic intermediate in animals, plants, and microorganisms. Phosphoenolpyruvic Acid Monopotassium Salt, MP Biomedicals is a phosphoric ester of the enol form of pyruvic acid. PEP is a high-energy compound involved in glycolysi

Phospho(enol)pyruvic acid (PEP) is involved in glycolysis and gluconeogeneis. In glycolysis, PEP is metabolized by Pyruvate Kinase to yield pyruvate. In plants, PEP is involved in the formation of aromatic amino acids as well as in the carbon fixation pathway.

PEP is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. PEP protects against the cell injury induced by oxidative stress and against the dysfunction in carbohydrate metabolism.

Specifications

CAS 4265-07-0
Melting Point 175°C (literature)(decomposition)
Molecular Formula C3H4KO6P
Quantity 5 g
Synonym potassium 1-carboxyvinyl hydrogenphosphate, potassium hydrogen 2-phosphonatooxy acrylate, 2-propenoic acid, 2-phosphonooxy-, monopotassium salt, potassium 1-carboxyvinyl hydrogen phosphate, phospho enol pyruvic acid monopotassium salt, phosphoenolpyruvic acid monopotassium salt, 2-propenoic acid, 2-phosphonooxy-, potassium salt 1:1, potassium 2-hydrogen phosphonooxy prop-2-enoic acid, phosphoenolpyruvate potassium salt, potassium 2-hydrogen phosphonatooxy prop-2-enoic acid
Solubility Information Soluble in water (100mg/mL)
InChI Key SOSDSEAIODNVPX-UHFFFAOYSA-M
SMILES C=C(C(=O)O)OP(=O)(O)[O-].[K+]
IUPAC Name potassium;1-carboxyethenyl hydrogen phosphate
Molecular Weight (g/mol) 206.131
PubChem CID 23678879
Formula Weight 206.1g/mol
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.