accessibility menu, dialog, popup

UserName

NaNaONOOPOOO

CAS RN 4264-83-9

IUPAC Name: disodium 4-nitrophenyl phosphate
Synonyms: disodium p-nitrophenyl phosphate
Molecular Weight (g/mol): 263.05
Molecular Formula: C6H4NNa2O6P
InChi Key: VIYFPAMJCJLZKD-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
Physical Form 
  • (5)
  • (1)
  • (4)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (6)
  • (2)
  • (2)
  • (5)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)