missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl Phosphate, Disodium Salt Hexahydrate, MP Biomedicals

Product Code. 11394069 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394069 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11394069 Supplier MP Biomedicals Supplier No. 02100878.5
  • ONLINE EXCLUSIVE PRICE
    43.00 EUR
    40.95 EUR / 500mg
    Save 2.05 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • p-nitrophenol is suitable for use as a substrate for alkaline and acid phosphatase

Specifications

CAS 4264-83-9
Melting Point >300°C (literature)
Boiling Point 457.84°C (760 mmHg(literature))
Molecular Formula C6H4NNa2O6P
Linear Formula C6H4NO6PNa2 . 6H2O
MDL Number MFCD00007319
Quantity 500 mg
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
Solubility Information Soluble in water.
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
IUPAC Name disodium 4-nitrophenyl phosphate
Molecular Weight (g/mol) 263.05
PubChem CID 77949
Formula Weight 371.14
Physical Form Powder
Chemical Name or Material p-NITROPHENYL PHOSPHATE, DISODIUM SALT HEXAHYDRATE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.