accessibility menu, dialog, popup

UserName

CH3NOPHOOHOOOH

CAS RN 41468-25-1

IUPAC Name: [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid hydrate
Synonyms: pyridoxal phosphate hydrate
Molecular Weight (g/mol): 265.16
Molecular Formula: C8H12NO7P
InChi Key: CEEQUQSGVRRXQI-UHFFFAOYSA-N
SMILES: O.CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (4)
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (9)
  • (3)
  • (4)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)