missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal-5-Phosphate Monohydrate, Approx. 98%, MP Biomedicals™

Product Code. 11426142 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426142 5 g 5g
11416142 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426142 Supplier MP Biomedicals Supplier No. 0210276905
  • ONLINE EXCLUSIVE PRICE
    128.00 EUR
    121.80 EUR / 5g
    Save 6.20 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Coenzyme

Chemical Identifiers

CAS 41468-25-1
Molecular Formula C8H10NO6P
Molecular Weight (g/mol) 247.14
MDL Number MFCD00006333
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
Synonym 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate
PubChem CID 38882
IUPAC Name [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Specifications

Color White
Assay Percent Range appox. 98%
Quantity 5 g
Formula Weight 265.1
Physical Form Solid
Chemical Name or Material Pyridoxal-5-Phosphate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.