missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal-5-Phosphate Monohydrate, MP Biomedicals™

Product Code. 11436142 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436142 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11436142 Supplier MP Biomedicals Supplier No. 0210276925
  • ONLINE EXCLUSIVE PRICE
    519.00 EUR
    493.50 EUR / 25g
    Save 25.50 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acts as a coenzyme in transamination, decarboxylation and deamination reactions. Pyridoxal-5-Phosphate Monohydrate, MP Biomedicals is an active form of Vitamin B6, an enzyme prosthetic group. It is useful for modifying lysyl and valyl residues in proteins

PLP acts as a coenzyme in conversion reactions, such as the conversion of dopa into dopamine, of glutamate into GABA, and of histidine to histamine. It works mechanistically through formation of a Schiff-base.

Specifications

CAS 41468-25-1
Melting Point 140°C to 143°C (literature)
Molecular Formula C8H10NO6P
MDL Number MFCD00006333
Quantity 25 g
Synonym 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate
Solubility Information Soluble in 1M Hydrochloric acid (50mg/mL; clear and colorless to light yellow-green solution).
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O
IUPAC Name [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
Molecular Weight (g/mol) 247.14
PubChem CID 38882
Formula Weight 265.1
Percent Purity ∼98%
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.