CAS RN 403640-27-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Methiozolin, TRC
CAS: 403640-27-7 Molecular Formula: C17 H17 F2 N O2 S Molecular Weight (g/mol): 337.3842 Synonym: Isoxazole, 5-[[(2,6-difluorophenyl)methoxy]methyl]-4,5-dihydro-5-methyl-3-(3-methyl-2-thienyl)-,5-[[(2,6-Difluorophenyl)methoxy]methyl]-4,5-dihydro-5-methyl-3-(3-methyl-2-thienyl)isoxazole,Methiozolin IUPAC Name: 5-[(2,6-difluorophenyl)methoxymethyl]-5-methyl-3-(3-methylthiophen-2-yl)-4H-1,2-oxazole SMILES: Cc1ccsc1C2=NOC(C)(COCc3c(F)cccc3F)C2
2
Methiozolin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality