missing translation for 'onlineSavingsMsg'
Learn More

Methiozolin, TRC

Product Code. 30180178
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180178 25 mg 25mg
30180123 5 mg 5mg
30180073 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180178 Supplier Toronto Research Chemicals Supplier No. TRCM77850525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403640-27-7
Molecular Formula C17 H17 F2 N O2 S
Molecular Weight (g/mol) 337.3842
Synonym Isoxazole, 5-[[(2,6-difluorophenyl)methoxy]methyl]-4,5-dihydro-5-methyl-3-(3-methyl-2-thienyl)-, 5-[[(2,6-Difluorophenyl)methoxy]methyl]-4,5-dihydro-5-methyl-3-(3-methyl-2-thienyl)isoxazole, Methiozolin
IUPAC Name 5-[(2,6-difluorophenyl)methoxymethyl]-5-methyl-3-(3-methylthiophen-2-yl)-4H-1,2-oxazole
SMILES Cc1ccsc1C2=NOC(C)(COCc3c(F)cccc3F)C2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H17F2NO2S/c1-11-6-7-23-16(11)15-8-17(2,22-20-15)10-21-9-12-13(18)4-3-5-14(12)19/h3-7H,8-10H2,1-2H3
Formula Weight 337.0948
Chemical Name or Material Methiozolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.