missing translation for 'onlineSavingsMsg'
Learn More

Methiozolin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30320179
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320179 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320179 Supplier Dr. Ehrenstorfer Supplier No. DREA15025000AL100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methiozolin
CAS 403640-27-7
Analyte or Component Names Methiozolin
Shipping Condition Room Temperature
Molecular Formula C17 H17 F2 N O2 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Isoxazole, 5-[[(2,6-difluorophenyl)methoxy]methyl]-4,5-dihydro-5-methyl-3-(3-methyl-2-thienyl)-, 5-[[(2,6-Difluorophenyl)methoxy]methyl]-4,5-dihydro-5-methyl-3-(3-methyl-2-thienyl)isoxazole
InChI Formula InChI=1S/C17H17F2NO2S/c1-11-6-7-23-16(11)15-8-17(2,22-20-15)10-21-9-12-13(18)4-3-5-14(12)19/h3-7H,8-10H2,1-2H3
SMILES Cc1ccsc1C2=NOC(C)(COCc3c(F)cccc3F)C2
IUPAC Name 5-[(2,6-difluorophenyl)methoxymethyl]-5-methyl-3-(3-methylthiophen-2-yl)-4H-1,2-oxazole
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 337.3842
Quantity 1 mL
Formula Weight 337.0948 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.