accessibility menu, dialog, popup

UserName

NH2BrFFF

CAS RN 393-36-2

IUPAC Name: 4-bromo-3-(trifluoromethyl)aniline
Synonyms: 4-bromo-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 240.02
Molecular Formula: C7H5BrF3N
InChi Key: YGNISOAUPSJDJE-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)