Learn More
4-Bromo-3-(trifluoromethyl)aniline, 98%, Thermo Scientific™
Product Code. 11468307
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468307 25 g 25g
11448307 1 g 1g
11458307 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11468307 Supplier Thermo Scientific Alfa Aesar Supplier No. A18380.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 393-36-2
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
MDL Number MFCD00007827
InChI Key YGNISOAUPSJDJE-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl aniline, 5-amino-2-bromobenzotrifluoride, 4-bromo-3-trifluoromethyl-phenylamine, 3-trifluoromethyl-4-bromoaniline, 4-amino-1-bromo-2-trifluoromethyl benzene, 4-bromo-alpha,alpha,alpha-trifluoro-m-toluidine, benzenamine, 4-bromo-3-trifluoromethyl, 5-amino-2-bromo benzotrifluoride, buttpark 37\11-68
PubChem CID 67852
IUPAC Name 4-bromo-3-(trifluoromethyl)aniline
SMILES NC1=CC=C(Br)C(=C1)C(F)(F)F
Specifications
Melting Point 46°C to 49°C
Boiling Point 81°C to 84°C (0.5 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN2811
Beilstein 641589
Formula Weight 240.03
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)aniline