Learn More
4-Bromo-3-(trifluoromethyl)aniline, TRC
Product Code. 30332314
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332314 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30332314 Supplier Toronto Research Chemicals Supplier No. TRCB810695250MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)aniline
CAS 393-36-2
Molecular Formula C7H5BrF3N
Quantity 250 mg
Synonym Bicalutamide Impurity 6
InChI Formula InChI=1S/C7H5BrF3N/c8-6-2-1-4(12)3-5(6)7(9,10)11/h1-3H,12H2
SMILES Nc1ccc(Br)c(c1)C(F)(F)F
IUPAC Name 4-bromo-3-(trifluoromethyl)aniline
Molecular Weight (g/mol) 240.02
Formula Weight 238.9557