CAS RN 375-22-4
Heptafluorobutyric acid, 99%
CAS: 375-22-4 Molecular Formula: C4HF7O2 Molecular Weight (g/mol): 214.04 MDL Number: MFCD00004171 InChI Key: YPJUNDFVDDCYIH-UHFFFAOYSA-N Synonym: heptafluorobutyric acid,perfluorobutyric acid,heptafluorobutanoic acid,perfluorobutanoic acid,butanoic acid, heptafluoro,heptafluoro-1-butanoic acid,perfluoropropanecarboxylic acid,butyric acid, heptafluoro,kyselina heptafluormaselna,kyselina heptafluormaselna czech PubChem CID: 9777 ChEBI: CHEBI:39426 IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutanoic acid SMILES: C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
Heptafluorobutyric acid, 99%
CAS: 375-22-4 Molecular Formula: C4HF7O2 Molecular Weight (g/mol): 214.039 MDL Number: MFCD00004171 InChI Key: YPJUNDFVDDCYIH-UHFFFAOYSA-N Synonym: heptafluorobutyric acid,perfluorobutyric acid,heptafluorobutanoic acid,perfluorobutanoic acid,butanoic acid, heptafluoro,heptafluoro-1-butanoic acid,perfluoropropanecarboxylic acid,butyric acid, heptafluoro,kyselina heptafluormaselna,kyselina heptafluormaselna czech PubChem CID: 9777 ChEBI: CHEBI:39426 IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutanoic acid SMILES: C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
Heptafluorobutyric Acid, 99.6%, MP Biomedicals™
CAS: 375-22-4 Molecular Formula: C4HF7O2 Molecular Weight (g/mol): 214.039 InChI Key: YPJUNDFVDDCYIH-UHFFFAOYSA-N Synonym: heptafluorobutyric acid,perfluorobutyric acid,heptafluorobutanoic acid,perfluorobutanoic acid,butanoic acid, heptafluoro,heptafluoro-1-butanoic acid,perfluoropropanecarboxylic acid,butyric acid, heptafluoro,kyselina heptafluormaselna,kyselina heptafluormaselna czech PubChem CID: 9777 ChEBI: CHEBI:39426 IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutanoic acid SMILES: C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
PFCA (C4-C14) Native Mixture 2 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Perfluorobutanoic acid 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Heptafluorobutyric Acid, TRC
CAS: 375-22-4 Molecular Formula: C4 H F7 O2 Molecular Weight (g/mol): 214.04 Synonym: Butanoic acid, heptafluoro- (9CI),Butyric acid, heptafluoro- (6CI,8CI),2,2,3,3,4,4,4-Heptafluorobutanoic acid,FC 23,Fluorad FC 23,H 0024,HFBA,Heptafluorobutanoic acid,Heptafluorobutyric acid,NSC 820,Perfluorobutanoic acid,Perfluorobutyric acid,Perfluoropropanecarboxylic acid,Butanoic acid, 2,2,3,3,4,4,4-heptafluoro-,PFBA IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutanoic acid SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)F
Heptafluorobutyric acid, 99%, for protein sequence analysis
CAS: 375-22-4 Molecular Formula: C4HF7O2 Molecular Weight (g/mol): 214.04 MDL Number: MFCD00004171 InChI Key: YPJUNDFVDDCYIH-UHFFFAOYSA-N Synonym: heptafluorobutyric acid,perfluorobutyric acid,heptafluorobutanoic acid,perfluorobutanoic acid,butanoic acid, heptafluoro,heptafluoro-1-butanoic acid,perfluoropropanecarboxylic acid,butyric acid, heptafluoro,kyselina heptafluormaselna,kyselina heptafluormaselna czech PubChem CID: 9777 ChEBI: CHEBI:39426 IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutanoic acid SMILES: C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O