missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric Acid Anhydrous, MP Biomedicals™

Product Code. 11403791 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403791 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11403791 Supplier MP Biomedicals Supplier No. 0215123780
  • ONLINE EXCLUSIVE PRICE
    372.00 EUR
    353.85 EUR / 100mL
    Save 18.15 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Heptafluorobutyric Acid is used in determination of amino acid sequences in proteins.

  • Heptafluorobutyric Acid is used as a mobile phase modifier which significantly improved selectivity in the HPLC analysis of histone proteins.
  • It can be used at a concentration of 0.1% in the mobile phase of an HPLC/LC-MS protocol for the detection of marine bacterioplankton siderophores.
  • It is used in the chiral LC analysis of (+)- and (-)-epibatidine.
  • The analysis of selenium containing compounds using HFBA in the MS matrix has been described.

Specifications

CAS 375-22-4
Density 1.645 g/mL
Molecular Formula C4HF7O2
Quantity 100 mL
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
Molecular Weight (g/mol) 214.039
PubChem CID 9777
ChEBI CHEBI:39426
Formula Weight 214
Chemical Name or Material Heptafluorobutyric Acid, Anhydrous
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.