CAS RN 3652-82-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Iodoethane-1,1-d2, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Iodoethane-1,1-d2 (Stabilized with Copper), TRC
CAS: 3652-82-2 Molecular Formula: C2 D2 H3 I Molecular Weight (g/mol): 157.98 Synonym: Ethyl-1,1-d2 Iodide,Ethyl Iodide-1,1-d2 IUPAC Name: 1,1-dideuterio-1-iodoethane SMILES: [2H]C([2H])(C)I