missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane-1,1-d2 (Stabilized with Copper), TRC

Product Code. 30403885
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403885 100 mg
30392030 50 mg
30399055 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30403885 Supplier Toronto Research Chemicals Supplier No. TRCI705874100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3652-82-2
Molecular Formula C2 D2 H3 I
Molecular Weight (g/mol) 157.98
Synonym Ethyl-1,1-d2 Iodide, Ethyl Iodide-1,1-d2
IUPAC Name 1,1-dideuterio-1-iodoethane
SMILES [2H]C([2H])(C)I

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C2H5I/c1-2-3/h2H2,1H3/i2D2
Formula Weight 157.9561
Chemical Name or Material Iodoethane-1,1-d2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.