Learn More
Iodoethane-1,1-d2, CDN
Product Code. 30311631
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311631 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30311631 Supplier CDN Supplier No. CDND00631G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Iodoethane-1,1-d2
CAS 3652-82-2
Molecular Formula C2 D2 H3 I
Quantity 1 g
Synonym Ethyl-1,1-d2 Iodide, Ethyl Iodide-1,1-d2
InChI Formula InChI=1 S/C2H5I/c1-2-3/h2H2,1H3/i2D2
SMILES [2 H]C([2 H])(C)I
IUPAC Name 1,1-dideuterio-1-iodoethane
Molecular Weight (g/mol) 157.98
Formula Weight 157.9561 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity