accessibility menu, dialog, popup

UserName

CAS RN 353229-59-1

CH3OONNSOOFFFHCl

CAS RN 353229-59-1

IUPAC Name: 2,2,2-trifluoro-N-[4-(2-methoxyphenoxy)phenyl]-N-[(pyridin-3-yl)methyl]ethane-1-sulfonamide hydrochloride
Synonyms: 2,2,2-trifluoro-N-[4-(2-methoxyphenoxy)phenyl]-N-(pyridin-3-ylmethyl)ethanesulfonamide hydrochloride
Molecular Weight (g/mol): 488.91
Molecular Formula: C21H20ClF3N2O4S
InChi Key: LPWFRDWTOKLHJC-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=CC=C1OC1=CC=C(C=C1)N(CC1=CN=CC=C1)S(=O)(=O)CC(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results