missing translation for 'onlineSavingsMsg'
Learn More

LY 487379 Hydrochloride, TRC

Product Code. 30483625
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483625 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483625 Supplier Toronto Research Chemicals Supplier No. TRCL48679010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,2-trifluoro-N-[4-(2-methoxyphenoxy)phenyl]-N-(pyridin-3-ylmethyl)ethanesulfonamide Hydrochloride
CAS 353229-59-1
Molecular Formula C21 H19 F3 N2 O4 S . Cl H
Quantity 10 mg
Synonym LY 487379 Hydrochloride
InChI Formula InChI=1S/C21H19F3N2O4S.ClH/c1-29-19-6-2-3-7-20(19)30-18-10-8-17(9-11-18)26(14-16-5-4-12-25-13-16)31(27,28)15-21(22,23)24;/h2-13H,14-15H2,1H3;1H
SMILES Cl.COc1ccccc1Oc2ccc(cc2)N(Cc3cccnc3)S(=O)(=O)CC(F)(F)F
IUPAC Name 2,2,2-trifluoro-N-[4-(2-methoxyphenoxy)phenyl]-N-(pyridin-3-ylmethyl)ethanesulfonamide;hydrochloride
Molecular Weight (g/mol) 488.908
Formula Weight 488.078
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.