CAS RN 3296-90-0
IUPAC Name:
2,2-bis(bromomethyl)propane-1,3-diol
Synonyms:
2,2-bis(bromomethyl)propane-1,3-diol
Molecular Weight (g/mol):
261.94
Molecular Formula:
C5H10Br2O2
InChi Key:
CHUGKEQJSLOLHL-UHFFFAOYSA-N
SMILES:
OCC(CO)(CBr)CBr
1
–
2
of
2
results
1
2,2-Bis(bromomethyl)-1,3-propanediol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Pentaerythritol Dibromide, TRC
CAS: 3296-90-0 Molecular Formula: C5 H10 Br2 O2 Molecular Weight (g/mol): 261.94 Synonym: 2,2-Bis(bromomethyl)-1,3-propanediol,1,3-Dibromo-2,2-bis(hydroxymethyl)propane,1,3-Dibromo-2,2-dihydroxymethylpropane,1,3-Dibromo-2,2-dimethylolpropane,2,2-Dibromomethyl-1,3-propanediol,Dibromoneopentyl glycol,FR 1138,FR 522,NSC 9001,Pentaerythritol dibromide,Pentaerythritol dibromohydrin IUPAC Name: 2,2-bis(bromomethyl)propane-1,3-diol SMILES: OCC(CO)(CBr)CBr