missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol Dibromide, TRC

Product Code. 30135537
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135537 1 g 1g
30132518 25 g 25g
30011979 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135537 Supplier Toronto Research Chemicals Supplier No. TRCP2699701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3296-90-0
Molecular Formula C5 H10 Br2 O2
Molecular Weight (g/mol) 261.94
Synonym 2,2-Bis(bromomethyl)-1,3-propanediol, 1,3-Dibromo-2,2-bis(hydroxymethyl)propane, 1,3-Dibromo-2,2-dihydroxymethylpropane, 1,3-Dibromo-2,2-dimethylolpropane, 2,2-Dibromomethyl-1,3-propanediol, Dibromoneopentyl glycol, FR 1138, FR 522, NSC 9001, Pentaerythritol dibromide, Pentaerythritol dibromohydrin
IUPAC Name 2,2-bis(bromomethyl)propane-1,3-diol
SMILES OCC(CO)(CBr)CBr

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H10Br2O2/c6-1-5(2-7,3-8)4-9/h8-9H,1-4H2
Formula Weight 259.9048
Percent Purity >95%
Chemical Name or Material Pentaerythritol Dibromide
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.