missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(bromomethyl)-1,3-propanediol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Pentaerythritol Dibromide |
| CAS | 3296-90-0 |
| Analyte or Component Names | Pentaerythritol Dibromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H10 Br2 O2 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 2,2-Bis(bromomethyl)-1,3-propanediol, 1,3-Dibromo-2,2-bis(hydroxymethyl)propane, 1,3-Dibromo-2,2-dihydroxymethylpropane, 1,3-Dibromo-2,2-dimethylolpropane, 2,2-Dibromomethyl-1,3-propanediol, Dibromoneopentyl glycol, FR 1138, FR 522, NSC 9001, Pentaerythritol dibromide, Pentaerythritol dibromohydrin |
| InChI Formula | InChI=1S/C5H10Br2O2/c6-1-5(2-7,3-8)4-9/h8-9H,1-4H2 |
| Solution Type | Neat |
| SMILES | OCC(CO)(CBr)CBr |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?