accessibility menu, dialog, popup

UserName

FSOO

CAS RN 329-98-6

IUPAC Name: phenylmethanesulfonyl fluoride
Synonyms: phenylmethylsulfonyl fluoride PMSF
Molecular Weight (g/mol): 174.19
Molecular Formula: C7H7FO2S
InChi Key: YBYRMVIVWMBXKQ-UHFFFAOYSA-N
SMILES: FS(=O)(=O)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (10)
  • (9)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (7)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (6)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (10)
  • (9)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (7)
  • (2)
  • (2)