missing translation for 'onlineSavingsMsg'
Learn More

Phenylmethylsulfonyl Fluoride, MP Biomedicals™

Product Code. 11495352 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495352 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11495352 Supplier MP Biomedicals Supplier No. 02195381.2
  • ONLINE EXCLUSIVE PRICE
    30.90 EUR
    29.40 EUR / 250mg
    Save 1.50 EUR
    4% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Irreversibly inhibits serine proteases, such as trypsin and chymotrypsin. Phenylmethylsulfonyl Fluoride, MP Biomedicals also inhibits mammalian acetyl cholinesterase (but not those from fish sources) and cysteine proteases (reversible by reduced thiols).

Administration of PMSF produces analgesia unrelated to its anticholinesterase effect, and prolongs the analgesic effect of centrally administered β-endorphin..

Specifications

CAS 329-98-6
Melting Point 89°C to 99°C
Molecular Formula C7H7FO2S
Quantity 250 mg
Synonym phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride
Solubility Information Soluble in methanol and ethanol
InChI Key YBYRMVIVWMBXKQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)CS(=O)(=O)F
IUPAC Name phenylmethanesulfonyl fluoride
Molecular Weight (g/mol) 174.189
PubChem CID 4784
ChEBI CHEBI:8102
Formula Weight 174.2
Concentration or Composition (by Analyte or Components) Working Concentration: Recommended concentration is 15-175ug/mL.
Physical Form Crystals
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.