CAS RN 32795-47-4
CAS RN 32795-47-4
IUPAC Name:
(2Z)-but-2-enedioic acid; 2-methyl-4-phenyl-1,2,3,4-tetrahydroisoquinolin-8-amine
Synonyms:
maleic acid; nomifensine
Molecular Weight (g/mol):
354.41
Molecular Formula:
C20H22N2O4
InChi Key:
GEOCVSMCLVIOEV-BTJKTKAUNA-N
SMILES:
OC(=O)\C=C/C(O)=O.CN1CC(C2=CC=CC=C2)C2=CC=CC(N)=C2C1
1
–
3
of
3
results
2
Nomifensine Hydrogen Maleate, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Nomifensine maleate, MedChemExpress
MedChemExpress Nomifensine maleate is a selective inhibitor of dopamine uptake, used in adult attention deficit disorder.