missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nomifensine Hydrogen Maleate, LoGiCal
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Nomifensine Hydrogen Maleate |
| CAS | 32795-47-4 |
| Analyte or Component Names | Nomifensine Hydrogen Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H18 N2 . C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,2,3,4-Tetrahydro-2-methyl-4-phenyl-8-isoquinolinamine (2Z)-2-butenedioate (1:1), 1,2,3,4-Tetrahydro-2-methyl-4-phenyl-8-isoquinolinamine (Z)-2-butenedioate (1:1), (±)-Nomifensine hydrogen maleate, Alival, Alival (antidepressant), HOE 984, Hostalival, Merital, NSC 289114, Neurolene, Nomifensine hydrogen maleate, Nomifensine maleate, Psicronizer |
| InChI Formula | InChI=1S/C16H18N2.C4H4O4/c1-18-10-14(12-6-3-2-4-7-12)13-8-5-9-16(17)15(13)11-18;5-3(6)1-2-4(7)8/h2-9,14H,10-11,17H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| Solution Type | Neat |
| SMILES | CN1CC(c2ccccc2)c3cccc(N)c3C1.OC(=O)\C=C/C(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction