missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Nomifensine

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Nomifensine is a potent inhibitor of noradrenaline and dopamine uptake (Ki values are 4.7, 26 and 4000 nM for inhibition of noradrenaline, dopamine and 5-HT uptake respectively, in rat brain). Increases central dopamine levels in rats following systemic administration. Displays antidepressant activity.
Specifications
Specifications
| CAS | 32795-47-4 |
| Quantity | 50 mg |
| Molecular Formula | C20H22N2O4 |
| Formula Weight | 354.4 |
| Purity | >98% |
| Molecular Weight (g/mol) | 354.406 |
| Chemical Name or Material | Nomifensine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction