accessibility menu, dialog, popup

UserName

CAS RN 315694-89-4

NNOO(E/Z)(E/Z)

CAS RN 315694-89-4

IUPAC Name: 2-(2-phenylethenyl)-4-[(pyridin-3-yl)methylidene]-4,5-dihydro-1,3-oxazol-5-one
Synonyms: 2-(2-phenylethenyl)-4-(pyridin-3-ylmethylidene)-1,3-oxazol-5-one
Molecular Weight (g/mol): 276.3
Molecular Formula: C17H12N2O2
InChi Key: GFGMISOSPOPSHN-UHFFFAOYSA-N
SMILES: O=C1OC(C=CC2=CC=CC=C2)=NC1=CC1=CN=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results