Learn More
TC-DAPK 6, MedChemExpress
Product Code. 16472677
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16472677
Brand: Medchem Express HY15513
MedChemExpress TC-DAPK 6 is a potent, ATP-competitive, and highly selective DAPK inhibitor (IC50=69 and 225 nM against DAPK1 and DAPK3, respectively, with 10 μM ATP).
TC-DAPK 6 is a potent, ATP-competitive, and highly selective DAPK inhibitor (IC50=69 and 225 nM against DAPK1 and DAPK3, respectively, with 10 μM ATP).
Specifications
Chemical Name or Material TC-DAPK 6
CAS 315694-89-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H12N2O2
Quantity 50 mg
Solubility Information DMSO : 18.67 mg/mL (67.57 mM; Need ultrasonic and warming)
SMILES O=C(OC(/C=C/C1=CC=CC=C1)=N/2)C2=C/C3=CC=CN=C3
Molecular Weight (g/mol) 276.29
Formula Weight 276.29
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease