Learn More
Tocris Bioscience™ TC-DAPK 6
Product Code. 18064652
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18064652
Brand: Tocris Bioscience™ 4301/10
Potent and selective inhibitor of DAPK1
Potent and selective, ATP-competitive inhibitor of death-associated protein kinase 1 (DAPK1) (IC50 values are 69 and 225 nM for DAPK1 and DAPK3 respectively, when assayed with 10 μM ATP). Displays selectivity for DAPK1 over a range of 48 other kinases, including Abl, AMPK, Chk1, Met and Src (IC50 > 10 μM).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 315694-89-4
Molecular Formula C17H12N2O2
Specifications
Quantity 10 mg
Formula Weight 276.29
Purity >98%
Molecular Weight (g/mol) 276.295
Chemical Name or Material TC-DAPK 6