accessibility menu, dialog, popup

UserName

CAS RN 292621-45-5

BrFOH2N

CAS RN 292621-45-5

IUPAC Name: 4-bromo-2-fluorobenzamide
Synonyms: 4-bromo-2-fluorobenzamide
Molecular Weight (g/mol): 218.03
Molecular Formula: C7H5BrFNO
InChi Key: MJDRFCPNHLHNON-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C=C(Br)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results