missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzamide, 96%

Product Code. 15458987
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15458987 250 mg 250mg
15448987 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15458987

Brand: Thermo Scientific Alfa Aesar H52336.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 292621-45-5 | C7H5BrFNO | 218.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 292621-45-5
Molecular Formula C7H5BrFNO
Molecular Weight (g/mol) 218.025
MDL Number MFCD03094432
InChI Key MJDRFCPNHLHNON-UHFFFAOYSA-N
Synonym acmc-209h7t, 4-bromo-2-fluoro-benzamide, benzamide,4-bromo-2-fluoro, benzamide, 4-bromo-2-fluoro
PubChem CID 2773331
IUPAC Name 4-bromo-2-fluorobenzamide
SMILES C1=CC(=C(C=C1Br)F)C(=O)N

Specifications

Melting Point 151°C to 153°C
Quantity 250 mg
Formula Weight 218.03
Percent Purity 96%
Chemical Name or Material 4-Bromo-2-fluorobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.