missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzamide, TRC

Product Code. 30345928
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30345928 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30345928

Brand: Toronto Research Chemicals TRCB6803151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromo-2-fluorobenzamide
CAS 292621-45-5
Molecular Formula C7H5BrFNO
Quantity 1 g
InChI Formula InChI=1S/C7H5BrFNO/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H2,10,11)
SMILES NC(=O)c1ccc(Br)cc1F
IUPAC Name 4-bromo-2-fluorobenzamide
Molecular Weight (g/mol) 218.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.