CAS RN 27581-13-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,3,4,5-Tetrabromobenzoic Acid, TRC
CAS: 27581-13-1 Molecular Formula: C7H2Br4O2 Molecular Weight (g/mol): 437.71 Synonym: 2,3,4,5-Tetrabromobenzoic acid,2,3,4,5-Tetrabromobenzoic acid IUPAC Name: 2,3,4,5-tetrabromobenzoic acid SMILES: BrC1=C(Br)C(Br)=CC(C(O)=O)=C1Br