missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrabromobenzoic Acid, TRC

Product Code. 30137235
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137235 10 mg 10mg
30134653 5 mg 5mg
30004374 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137235 Supplier Toronto Research Chemicals Supplier No. TRCT29114010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27581-13-1
Molecular Formula C7H2Br4O2
Molecular Weight (g/mol) 437.71
Synonym 2,3,4,5-Tetrabromobenzoic acid
IUPAC Name 2,3,4,5-tetrabromobenzoic acid
SMILES BrC1=C(Br)C(Br)=CC(C(O)=O)=C1Br

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H2Br4O2/c8-3-1-2(7(12)13)4(9)6(11)5(3)10/h1H,(H,12,13)
Formula Weight 433.68
Chemical Name or Material 2,3,4,5-Tetrabromobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.