accessibility menu, dialog, popup

UserName

NN

CAS RN 2739-97-1

IUPAC Name: 2-(pyridin-2-yl)acetonitrile
Synonyms: 2-(pyridin-2-yl)acetonitrile
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: UKVQBONVSSLJBB-UHFFFAOYSA-N
SMILES: N#CCC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)