Learn More
2-Pyridineacetonitrile, 97%
Product Code. 11444414
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444414 100 g 100g
11424414 5 g 5g
11434414 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11444414 Supplier Thermo Scientific Alfa Aesar Supplier No. A13087.22
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
CAS: 2739-97-1 | C7H6N2 | 118.139 g/mol
2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2739-97-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00006346
InChI Key UKVQBONVSSLJBB-UHFFFAOYSA-N
Synonym 2-pyridylacetonitrile, 2-pyridineacetonitrile, pyridine-2-acetonitrile, 2-pyridin-2-yl acetonitrile, 2-2-pyridyl acetonitrile, 2-cyanomethyl pyridine, pyridine-acetonitrile, pyridyl-2-acetonitrile, pyridin-2-ylacetonitrile, pyridin-2-yl acetonitrile
PubChem CID 75959
IUPAC Name 2-pyridin-2-ylacetonitrile
SMILES C1=CC=NC(=C1)CC#N
Specifications
Melting Point 23°C to 25°C
Density 1.059
Boiling Point 66°C to 68°C (1 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.525
Quantity 100 g
UN Number UN3439
Beilstein 111215
Solubility Information Miscible with water.
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material 2-Pyridineacetonitrile