Learn More
2-Pyridineacetonitrile, TRC
Product Code. 30427445
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427445 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30427445 Supplier Toronto Research Chemicals Supplier No. TRCP996955500MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Pyridineacetonitrile
CAS 2739-97-1
Molecular Formula C7H6N2
Quantity 500 mg
Synonym (2-Pyridinyl)acetonitrile, (Pyridyl-2-yl)acetonitrile, 2-(2-Pyridyl)acetonitrile
InChI Formula InChI=1S/C7H6N2/c8-5-4-7-3-1-2-6-9-7/h1-3,6H,4H2
SMILES N#CCC1=CC=CC=N1
IUPAC Name 2-(pyridin-2-yl)acetonitrile
Molecular Weight (g/mol) 118.14
Formula Weight 118.05