accessibility menu, dialog, popup

UserName

CAS RN 2646-71-1

N(R)(R)(R)NN(R)OOPPPO(R)(R)(R)(S)ONNOOOOOOOOOOHONH2NH2HOOHNaNaNaNa

CAS RN 2646-71-1

IUPAC Name: {[(2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-[({[({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)methyl]-4-hydroxyoxolan-3-yl]oxy}phosphonic acid
Synonyms: nadph
Molecular Weight (g/mol): 745.42
Molecular Formula: C21H30N7O17P3
InChi Key: ACFIXJIJDZMPPO-NNYOXOHSSA-N
SMILES: NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP(O)(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Percent Purity 
  • (1)
  • (6)
  • (2)
  • (2)
  • (5)
  • (2)
Physical Form 
  • (14)
  • (4)
Quantity 
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
  • (2)
  • (2)
  • (5)
  • (2)
  • (14)
  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)