missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 98%, MP Biomedicals™
Shop All MP Biomedicals ProductsSpecifications
Specifications
| Color | White |
| Quantity | 250 mg |
| IUPAC Name | tetrasodium [(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonatooxy)oxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
| Formula Weight | 833.4 |
| Percent Purity | 0.98 |
| Physical Form | Powder |
| Chemical Name or Material | β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?