CAS RN 261763-31-9
IUPAC Name:
1-(2,6-difluoro-3-methylphenyl)ethan-1-one
Synonyms:
1-(2,6-difluoro-3-methylphenyl)ethanone
Molecular Weight (g/mol):
170.16
Molecular Formula:
C9H8F2O
InChi Key:
BLSXKYBXBXVYJZ-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(F)C=CC(C)=C1F
1
–
1
of
1
results
1
2',6'-Difluoro-3'-methylacetophenone, 97%, Thermo Scientific™
CAS: 261763-31-9 Molecular Formula: C9H8F2O Molecular Weight (g/mol): 170.16 MDL Number: MFCD01631339 InChI Key: BLSXKYBXBXVYJZ-UHFFFAOYSA-N Synonym: 2',6'-difluoro-3'-methylacetophenone,1-2,6-difluoro-3-methylphenyl ethanone,2,6-difluoro-3-methylacetophenone,1-acetyl-2,6-difluoro-3-methylbenzene,acmc-20al2p,1-2,6-difluoro-3-methyphenyl-ethanone,1-2,6-bis fluoranyl-3-methyl-phenyl ethanone,ethanone, 1-2,6-difluoro-3-methylphenyl-9ci PubChem CID: 2774132 IUPAC Name: 1-(2,6-difluoro-3-methylphenyl)ethanone SMILES: CC(=O)C1=C(F)C=CC(C)=C1F