CAS RN 26020-48-4
IUPAC Name:
trichlorothiophene-2-carboxylic acid
Synonyms:
trichlorothiophene-2-carboxylic acid
Molecular Weight (g/mol):
231.47
Molecular Formula:
C5HCl3O2S
InChi Key:
FVFYTPFJDMYLJS-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(Cl)C(Cl)=C(Cl)S1
1
–
1
of
1
results
1
Alfa Aesar™ 3,4,5-Trichlorothiophene-2-carboxylic acid, 98+%
CAS: 26020-48-4 Molecular Formula: C5HCl3O2S Molecular Weight (g/mol): 231.47 MDL Number: MFCD00092429 InChI Key: FVFYTPFJDMYLJS-UHFFFAOYSA-N Synonym: 3,4,5-trichloro-2-thiophenecarboxylic acid,trichlorothiophene-2-carboxylic acid,acmc-1cbhi,3,4,5-trichlorothio-phene-2-carboxylic acid,3,4,5-tris chloranyl thiophene-2-carboxylic acid PubChem CID: 4168584 SMILES: OC(=O)C1=C(Cl)C(Cl)=C(Cl)S1