missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3,4,5-Trichlorothiophene-2-carboxylic acid, 98+%
Description
Applications
It is a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
It is a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
Specifications
Specifications
| Melting Point | 226°C to 228°C |
| Quantity | 5 g |
| Beilstein | 155175 |
| Solubility Information | Insoluble in water. |
| IUPAC Name | 3,4,5-trichlorothiophene-2-carboxylic acid |
| Formula Weight | 231.49 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 3,4,5-Trichlorothiophene-2-carboxylic acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?