CAS RN 24796-94-9
IUPAC Name:
3,7-bis(diethylamino)-5λ⁴-phenoxazin-5-ylium perchlorate
Synonyms:
oxazine-1 perchlorate
Molecular Weight (g/mol):
423.89
Molecular Formula:
C20H26ClN3O5
InChi Key:
PKZWDLHLOBYXKV-UHFFFAOYSA-M
SMILES:
[O-][Cl](=O)(=O)=O.CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3N=C2C=C1)N(CC)CC
1
–
1
of
1
results
1
MedChemExpress Oxazine 1 perchlorate
MedChemExpress Oxazine 1 perchlorate is a symmetric cationic dye (λex=653 nm, λem=666 nm).