CAS RN 2396-84-1
CAS RN 2396-84-1
IUPAC Name:
ethyl (2E,4E)-hexa-2,4-dienoate
Synonyms:
ethyl (2E,4E)-hexa-2,4-dienoate
Molecular Weight (g/mol):
140.18
Molecular Formula:
C8H12O2
InChi Key:
OZZYKXXGCOLLLO-TWTPFVCWSA-N
SMILES:
CCOC(=O)\C=C\C=C\C
1
–
1
of
1
results
1
Ethyl sorbate, 96%
CAS: 2396-84-1 Molecular Formula: C8H12O2 Molecular Weight (g/mol): 140.18 MDL Number: MFCD00009296 InChI Key: OZZYKXXGCOLLLO-TWTPFVCWSA-N Synonym: ethyl sorbate,ethyl 2,4-hexadienoate,ethyl hexa-2,4-dienoate,sorbic acid, ethyl ester,2,4-hexadienoic acid, ethyl ester,ethyl 2e,4e-hexa-2,4-dienoate,ethyl e,e-2,4-hexadienoate,unii-hsr16usg4d,2,4-hexadienoic acid, ethyl ester, 2e,4e,fema no. 2459 PubChem CID: 1550470 IUPAC Name: ethyl (2E,4E)-hexa-2,4-dienoate SMILES: CCOC(=O)\C=C\C=C\C